MilliporeSigma
Caspr1 is a host receptor for meningitis-causing Escherichia coli.
Nature communications (2018-06-14)
Wei-Dong Zhao, Dong-Xin Liu, Jia-Yi Wei, Zi-Wei Miao, Ke Zhang, Zheng-Kang Su, Xue-Wei Zhang, Qiang Li, Wen-Gang Fang, Xiao-Xue Qin, De-Shu Shang, Bo Li, Qing-Chang Li, Liu Cao, Kwang Sik Kim, Yu-Hua Chen
ABSTRACT
Escherichia coli is the leading cause of neonatal Gram-negative bacterial meningitis, but the pathogenesis of E. coli meningitis remains elusive. E. coli penetration of the blood-brain barrier (BBB) is the critical step for development of meningitis. Here, we identify Caspr1, a single-pass transmembrane protein, as a host receptor for E. coli virulence factor IbeA to facilitate BBB penetration. Genetic ablation of endothelial Caspr1 and blocking IbeA-Caspr1 interaction effectively prevent E. coli penetration into the brain during meningitis in rodents. IbeA interacts with extracellular domain of Caspr1 to activate focal adhesion kinase signaling causing E. coli internalization into the brain endothelial cells of BBB. E. coli can invade hippocampal neurons causing apoptosis dependent on IbeA-Caspr1 interaction. Our results indicate that E. coli exploits Caspr1 as a host receptor for penetration of BBB resulting in meningitis, and that Caspr1 might be a useful target for prevention or therapy of E. coli meningitis.
Sigma-Aldrich
Anti-FAK Antibody, clone 4.47, clone 4.47, Upstate®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human CNTNAP1