MilliporeSigma
Neuregulin-1 attenuates stress-induced vascular senescence.
Cardiovascular research (2018-03-13)
Hadis Shakeri, Andreas B Gevaert, Dorien M Schrijvers, Guido R Y De Meyer, Gilles W De Keulenaer, Pieter-Jan D F Guns, Katrien Lemmens, Vincent F Segers
ABSTRACT
Cardiovascular ageing is a key determinant of life expectancy. Cellular senescence, a state of irreversible cell cycle arrest, is an important contributor to ageing due to the accumulation of damaged cells. Targeting cellular senescence could prevent age-related cardiovascular diseases. In this study, we investigated the effects of neuregulin-1 (NRG-1), an epidermal growth factor with cardioprotective and anti-atherosclerotic effects, on cellular senescence. Senescence was induced in cultured rat aortic endothelial cells (ECs) and aortic smooth muscle cells (SMCs) by 2 h exposure to 30 µM hydrogen peroxide (H2O2). Cellular senescence was confirmed after 72 h using senescence-associated-β-galactosidase staining (SA-β-gal), cell surface area, and western blot analyses of SA pathways (acetyl-p53, p21). Recombinant human NRG-1 (rhNRG-1, 20 ng/mL) significantly reduced H2O2-induced senescence, as shown by a lower number of SA-β-gal positive cells, smaller surface area and lower expression of acetyl-p53. In C57BL/6 male mice rendered diabetic with streptozotocin (STZ), rhNRG-1 attenuated cellular senescence in aortic ECs and SMCs. Next, we created mice with SMC-specific knockdown of the NRG-1 receptor ErbB4. Aortic SMCs isolated from SMC-specific ErbB4 deficient mice (ErbB4f/+ SM22α-Cre+) showed earlier cellular senescence in vitro compared with wild-type (ErbB4+/+ SM22α-Cre+) SMCs. Furthermore, when rendered diabetic with STZ, ErbB4f/+ SM22α-Cre+ male mice showed significantly more vascular senescence than their diabetic wild-type littermates and had increased mortality. This study is the first to explore the role of NRG-1 in vascular senescence. Our data demonstrate that NRG-1 markedly inhibits stress-induced premature senescence in vascular cells in vitro and in the aorta of diabetic mice in vivo. Consistently, deficiency in the NRG-1 receptor ErbB4 provokes cellular senescence in vitro as well as in vivo.
Sigma-Aldrich
Anti-Acetyl (Lys 319) p53 Antibody, C-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid