MilliporeSigma
  • Neurofibromin Deficiency Induces Endothelial Cell Proliferation and Retinal Neovascularization.
Neurofibromin Deficiency Induces Endothelial Cell Proliferation and Retinal Neovascularization.
Investigative ophthalmology & visual science (2018-05-31)
Hanfang Zhang, Farlyn Z Hudson, Zhimin Xu, Rebekah Tritz, Modesto Rojas, Chintan Patel, Stephen B Haigh, Zsuzsanna Bordán, David A Ingram, David J Fulton, Neal L Weintraub, Ruth B Caldwell, Brian K Stansfield
ABSTRACT
Neurofibromatosis type 1 (NF1) is the result of inherited mutations in the NF1 tumor suppressor gene, which encodes the protein neurofibromin. Eye manifestations are common in NF1 with recent reports describing a vascular dysplasia in the retina and choroid. Common features of NF1 retinopathy include tortuous and dilated feeder vessels that terminate in capillary tufts, increased endothelial permeability, and neovascularization. Given the retinal vascular phenotype observed in persons with NF1, we hypothesize that preserving neurofibromin may be a novel strategy to control pathologic retinal neovascularization. Nf1 expression in human endothelial cells (EC) was reduced using small hairpin (sh) RNA and EC proliferation, migration, and capacity to form vessel-like networks were assessed in response to VEGF and hypoxia. Wild-type (WT), Nf1 heterozygous (Nf1+/-), and Nf1flox/+;Tie2cre pups were subjected to hyperoxia/hypoxia using the oxygen-induced retinopathy model. Retinas were analyzed quantitatively for extent of retinal vessel dropout, neovascularization, and capillary branching. Neurofibromin expression was suppressed in response to VEGF, which corresponded with activation of Mek-Erk and PI3-K-Akt signaling. Neurofibromin-deficient EC exhibited enhanced proliferation and network formation in response to VEGF and hypoxia via an Akt-dependent mechanism. In response to hyperoxia/hypoxia, Nf1+/- retinas exhibited increased vessel dropout and neovascularization when compared with WT retinas. Neovascularization was similar between Nf1+/- and Nf1flox/+;Tie2cre retinas, but capillary drop out in Nf1flox/+;Tie2cre retinas was significantly reduced when compared with Nf1+/- retinas. These data suggest that neurofibromin expression is essential for controlling endothelial cell proliferation and retinal neovascularization and therapies targeting neurofibromin-deficient EC may be beneficial.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes