MilliporeSigma
  • Catalytic Defluoroalkylation of Trifluoromethylaromatics with Unactivated Alkenes.
Catalytic Defluoroalkylation of Trifluoromethylaromatics with Unactivated Alkenes.
Journal of the American Chemical Society (2017-12-20)
Hengbin Wang, Nathan T Jui
ABSTRACT
We describe a new catalytic approach to selective functionalization of the strong C-F bonds in trifluoromethylaromatic (Ar-CF3) systems. In this approach, single electron reduction of Ar-CF3 substrates (using a photoredox catalyst) results in difluorobenzylic radical formation through a C-F cleavage mechanism. These radicals undergo efficient intermolecular coupling with simple alkenes in a defluoroalkylation process where radical termination is accomplished by a polarity reversal catalyst. This mild catalytic protocol engages a wide range of substrates to give medicinally relevant fluorinated substructures with complete regiocontrol.
Sigma-Aldrich
10-Phenylphenothiazine, ≥95%