MilliporeSigma
  • The role of IGF-I and its binding proteins in the development of type 2 diabetes and cardiovascular disease.
The role of IGF-I and its binding proteins in the development of type 2 diabetes and cardiovascular disease.
Diabetes, obesity & metabolism (2008-02-14)
Vivienne A Ezzat, Edward R Duncan, Stephen B Wheatcroft, Mark T Kearney
ABSTRACT
Patients with insulin resistance and type 2 diabetes have an excessive risk of cardiovascular disease (CVD); this increased risk is not fully explained by traditional risk factors such as hypertension and dyslipidaemias. There is now compelling evidence to suggest that abnormalities of insulin-like growth factor-I (IGF-I) and one of its binding proteins, insulin-like growth factor-binding protein-1 (IGFBP-1), occur in insulin-resistant states and may be significant factors in the pathophysiology of CVD. We reviewed articles and relevant bibliographies following a systematic search of MEDLINE for English language articles between 1966 and the present, using an initial search strategy combining the MeSH terms: IGF, diabetes and CVD. Our aim was first to review the role of IGF-I in vascular homeostasis and to explore the mechanisms by which it may exert its effects. We also present an overview of the physiology of the IGF-binding proteins, and finally, we sought to summarize the evidence to date describing the changes in the insulin/IGF-I/IGFBP-1 axis that occur in type 2 diabetes and CVD; in particular, we have focused on the potential vasculoprotective effects of both IGF-I and IGFBP-1. We conclude that this system represents an interesting and novel therapeutic target in the prevention of CVD in type 2 diabetes.
Sigma-Aldrich
Interferon-λ human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IFN-LAMBDA1 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)