MilliporeSigma
  • Production of IL-10 by human natural killer cells stimulated with IL-2 and/or IL-12.
Production of IL-10 by human natural killer cells stimulated with IL-2 and/or IL-12.
Journal of immunology (Baltimore, Md. : 1950) (1998-03-24)
P T Mehrotra, R P Donnelly, S Wong, H Kanegane, A Geremew, H S Mostowski, K Furuke, J P Siegel, E T Bloom
ABSTRACT
Human NK cell activity can be augmented in vitro by stimulation with IL-2 or IL-12, both of which also induce the production of IFN-gamma, TNF-alpha, and granulocyte-macrophage CSF by NK cells. For the first time, we demonstrate that freshly purified NK cells stimulated with IL-2 proliferated and produced IL-10 in a dose-dependent manner. IL-10 mRNA expression, as detected by semiquantitative reverse transcription-PCR, reached peak levels at 24 h. IL-10 protein was detectable on day 2 and further increased on days 3 and 6 as measured by ELISA. However, IL-12 alone induced neither substantial proliferation nor detectable IL-10 production by fresh NK cells, but it synergized with IL-2 in inducing IL-10 mRNA expression and protein synthesis. IL-10 production by activated NK cells was confirmed by intracytoplasmic cytokine staining by three-color immunofluorescence of CD16+ and/or CD56+ NK cells with anti-IL-10 antibody. IL-10 production by NK cells was further confirmed in the NK-like cell line, YT, which constitutively expressed IL-10 mRNA and protein. IL-12 alone did not induce NK proliferation, but it inhibited IL-2-induced proliferation. Neutralization of endogenously produced IL-10 with anti-IL-10 antibodies did not overcome the inhibition of IL-2-induced proliferation by IL-12. Together, these results demonstrate that IL-2 and IL-12 synergize to induce IL-10 production by human NK cells and that IL-12 inhibits IL-2 induced NK cell proliferation by an IL-10-independent mechanism.
Sigma-Aldrich
IL-10 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
IP-10 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-10 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)