MilliporeSigma
  • Neuritin: a gene induced by neural activity and neurotrophins that promotes neuritogenesis.
Neuritin: a gene induced by neural activity and neurotrophins that promotes neuritogenesis.
Proceedings of the National Academy of Sciences of the United States of America (1997-03-18)
G S Naeve, M Ramakrishnan, R Kramer, D Hevroni, Y Citri, L E Theill
ABSTRACT
Neural activity and neurotrophins induce synaptic remodeling in part by altering gene expression. A cDNA encoding a glycosylphoshatidylinositol-anchored protein was identified by screening for hippocampal genes that are induced by neural activity. This molecule, named neuritin, is expressed in postmitotic-differentiating neurons of the developing nervous system and neuronal structures associated with plasticity in the adult. Neuritin message is induced by neuronal activity and by the activity-regulated neurotrophins BDNF and NT-3. Purified recombinant neuritin promotes neurite outgrowth and arborization in primary embryonic hippocampal and cortical cultures. These data implicate neuritin as a downstream effector of activity-induced neurite outgrowth.
Sigma-Aldrich
Neuritin human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture