MilliporeSigma
  • Optimization of a multiple headspace sorptive extraction method coupled to gas chromatography-mass spectrometry for the determination of volatile compounds in macroalgae.
Optimization of a multiple headspace sorptive extraction method coupled to gas chromatography-mass spectrometry for the determination of volatile compounds in macroalgae.
Journal of chromatography. A (2018-04-14)
Astrid Maruti, Enrique Durán-Guerrero, Carmelo G Barroso, Remedios Castro
ABSTRACT
A novel extraction technique is proposed in which the Multiple Headspace Extraction (MHE) approach is used in conjunction with Headspace Sorptive Extraction (HSSE) and Gas Chromatography-Mass Spectrometry (GC-MS) detection. The extraction method was developed to determine volatile compounds in macroalgae. Optimization of the extraction parameters was carried out using design of experiments to identify factors that affect the extraction: extraction time, temperature, twister length and amount of sample. The results of the optimization led to an extraction of 2 g of sample using a 20 mm Twister® at 66 °C for 180 min. The progression constants (β) were calculated for 43 volatile compounds, 29 of which could be quantified using the method. Linearity was attained with a determination coefficient higher than 0.99 for all studied compounds. Inter-day and inter-twister precisions ranged from 0.22% to 19.01% and from 0.69% to 14.76% respectively, and values below 10% were obtained for the majority of compounds. LOD and LOQ values ranged from the values obtained for diethyl succinate (0.012 μg/L and 0.088 μg/L, respectively) and those obtained for dimethyl sulfide (5.544 μg/L and 40.286 μg/L, respectively). However, for the majority of compounds values obtained were below 1 μg/L (LOD) and 5 μg/L (LOQ). Compounds such as ethyl acetate, hexanal, heptadecane, 2-hexenal, 6-methyl-5-hepten-2-one, dimethyl sulfide, benzyl alcohol, beta ionone, or beta cyclocitral, among others were correctly determined in three species of macroalgae: Ulva sp., Gracillaria sp. and Enteromorpha sp.
Sigma-Aldrich
2,4-Heptadienal, technical grade, 90%
Sigma-Aldrich
1-Chloropentane, 99%
Sigma-Aldrich
Isododecane, mixture of isomers, technical, ≥80% (GC)