MilliporeSigma
MUB40 Binds to Lactoferrin and Stands as a Specific Neutrophil Marker.
Cell chemical biology (2018-02-27)
Mark C Anderson, Thibault Chaze, Yves-Marie Coïc, Louise Injarabian, Friederike Jonsson, Naelle Lombion, Dorothée Selimoglu-Buet, Judith Souphron, Caroline Ridley, Pascale Vonaesch, Bruno Baron, Ellen T Arena, Jean-Yves Tinevez, Giulia Nigro, Katharina Nothelfer, Eric Solary, Valérie Lapierre, Thierry Lazure, Mariette Matondo, David Thornton, Philippe J Sansonetti, Françoise Baleux, Benoit S Marteyn
ABSTRACT
Neutrophils represent the most abundant immune cells recruited to inflamed tissues. A lack of dedicated tools has hampered their detection and study. We show that a synthesized peptide, MUB40, binds to lactoferrin, the most abundant protein stored in neutrophil-specific and tertiary granules. Lactoferrin is specifically produced by neutrophils among other leukocytes, making MUB40 a specific neutrophil marker. Naive mammalian neutrophils (human, guinea pig, mouse, rabbit) were labeled by fluorescent MUB40 conjugates (-Cy5, Dylight405). A peptidase-resistant retro-inverso MUB40 (RI-MUB40) was synthesized and its lactoferrin-binding property validated. Neutrophil lactoferrin secretion during in vitro Shigella infection was assessed with RI-MUB40-Cy5 using live cell microscopy. Systemically administered RI-MUB40-Cy5 accumulated at sites of inflammation in a mouse arthritis inflammation model in vivo and showed usefulness as a potential tool for inflammation detection using non-invasive imaging. Improving neutrophil detection with the universal and specific MUB40 marker will aid the study of broad ranges of inflammatory diseases.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Maleimide, 99%