MilliporeSigma
  • Tat controls transcriptional persistence of unintegrated HIV genome in primary human macrophages.
Tat controls transcriptional persistence of unintegrated HIV genome in primary human macrophages.
Virology (2018-03-20)
Beatrix Meltzer, Deemah Dabbagh, Jia Guo, Fatah Kashanchi, Mudit Tyagi, Yuntao Wu
ABSTRACT
In HIV infected macrophages, a large population of viral genomes persists as the unintegrated form (uDNA) that is transcriptionally active. However, how this transcriptional activity is controlled remains unclear. In this report, we investigated whether Tat, the viral transactivator of transcription, is involved in uDNA transcription. We demonstrate that de novo Tat activity is generated from uDNA, and this uDNA-derived Tat (uTat) transactivates the uDNA LTR. In addition, uTat is required for the transcriptional persistence of uDNA that is assembled into repressive episomal minichromatin. In the absence of uTat, uDNA minichromatin is gradually silenced, but remains highly inducible by HDAC inhibitors (HDACi). Therefore, functionally, uTat antagonizes uDNA minichromatin repression to maintain persistent viral transcription in macrophages. uTat-mediated viral persistence may establish a viral reservoir in macrophages where uDNA were found to persist.
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial