MilliporeSigma
  • Synthesis of a Novel Type of 2,3'-BIMs via Platinum-Catalysed Reaction of Indolylallenes with Indoles.
Synthesis of a Novel Type of 2,3'-BIMs via Platinum-Catalysed Reaction of Indolylallenes with Indoles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2018-02-03)
Lisa Cooper, José Miguel Alonso, Louise Eagling, Helen Newson, Sachini Herath, Christopher Thomson, Andrew Lister, Catherine Howsham, Brian Cox, María Paz Muñoz
ABSTRACT
Optimisation, scope and mechanism of the platinum-catalysed addition of indoles to indolylallenes is reported here to give 2,3'-BIMs with a novel core structure very relevant for pharmaceutical industry. The reaction is modulated by the electronic properties of the substituents on both indoles, with the 2,3'-BIMs favoured when electron donating groups are present. Although simple at first, a complex mechanism has been uncovered that explains the different behaviour of these systems with platinum when compared with other metals (e.g. gold). Detailed labelling studies have shown Pt-catalysed 6-endo-trig cyclisation of the indollylallene as the first step of the reaction and the involvement of two cyclic vinyl-platinum intermediates in equilibrium through a platinum carbene, as the key intermediates of the catalytic cycle towards the second nucleophilic attack and formation of the BIMs.
Sigma-Aldrich
5-Bromoindole, 99%
Sigma-Aldrich
5-Methoxyindole, 99%