MilliporeSigma
  • Competitive Adsorption between Nanoparticles and Surface Active Ions for the Oil-Water Interface.
Competitive Adsorption between Nanoparticles and Surface Active Ions for the Oil-Water Interface.
Langmuir : the ACS journal of surfaces and colloids (2018-04-11)
Xiaoqing Hua, Michael A Bevan, Joelle Frechette
ABSTRACT
Nanoparticles (NPs) can add functionality (e.g., catalytic, optical, rheological) to an oil-water interface. Adsorption of ∼10 nm NPs can be reversible; however, the mechanisms for adsorption and its effects on surface pressure remain poorly understood. Here we demonstrate how the competitive reversible adsorption of NPs and surfactants at fluid interfaces can lead to independent control of both the adsorbed amount and surface pressure. In contrast to prior work, both species investigated (NPs and surfactants) interact reversibly with the interface and without the surface active species binding to NPs. Independent measurements of the adsorption and surface pressure isotherms allow determination of the equation of state (EOS) of the interface under conditions where the NPs and surfactants are both in dynamic equilibrium with the bulk phase. The adsorption and surface pressure measurements are performed with gold NPs of two different sizes (5 and 10 nm), at two pH values, and across a wide concentration range of surfactant (tetrapentylammonium, TPeA+) and NPs. We show that free surface active ions compete with NPs for the interface and give rise to larger surface pressures upon the adsorption of NPs. Through a competitive adsorption model, we decouple the contributions of NPs wetting at the interface and their surface activity on the measured surface pressure. We also demonstrate reversible control of adsorbed amount via changes in the surfactant concentration or the aqueous phase pH.
Sigma-Aldrich
Tetrapentylammonium hydroxide solution, ~20% in H2O (T)
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%