MilliporeSigma
  • A systems biology approach to investigate the response of Synechocystis sp. PCC6803 to a high salt environment.
A systems biology approach to investigate the response of Synechocystis sp. PCC6803 to a high salt environment.
Saline systems (2009-09-09)
Jagroop Pandhal, Josselin Noirel, Phillip C Wright, Catherine A Biggs
ABSTRACT
Salt overloading during agricultural processes is causing a decrease in crop productivity due to saline sensitivity. Salt tolerant cyanobacteria share many cellular characteristics with higher plants and therefore make ideal model systems for studying salinity stress. Here, the response of fully adapted Synechocystis sp. PCC6803 cells to the addition of 6% w/v NaCl was investigated using proteomics combined with targeted analysis of transcripts. Isobaric mass tagging of peptides led to accurate relative quantitation and identification of 378 proteins, and approximately 40% of these were differentially expressed after incubation in BG-11 media supplemented with 6% salt for 9 days. Protein abundance changes were related to essential cellular functional alterations. Differentially expressed proteins involved in metabolic responses were also analysed using the probabilitistic tool Mixed Model on Graphs (MMG), where the role of energy conversion through glycolysis and reducing power through pentose phosphate pathway were highlighted. Temporal RT-qPCR experiments were also run to investigate protein expression changes at the transcript level, for 14 non-metabolic proteins. In 9 out of 14 cases the mRNA changes were in accordance with the proteins. Synechocystis sp. PCC6803 has the ability to regulate essential metabolic processes to enable survival in high salt environments. This adaptation strategy is assisted by further regulation of proteins involved in non-metabolic cellular processes, supported by transcriptional and post-transcriptional control. This study demonstrates the effectiveness of using a systems biology approach in answering environmental, and in particular, salt adaptation questions in Synechocystis sp. PCC6803.
Sigma-Aldrich
Cyanobacteria BG-11 Freshwater Solution, 50 ×, liquid, suitable for plant cell culture