MilliporeSigma
  • A novel small molecule chaperone of rod opsin and its potential therapy for retinal degeneration.
A novel small molecule chaperone of rod opsin and its potential therapy for retinal degeneration.
Nature communications (2018-05-19)
Yuanyuan Chen, Yu Chen, Beata Jastrzebska, Marcin Golczak, Sahil Gulati, Hong Tang, William Seibel, Xiaoyu Li, Hui Jin, Yong Han, Songqi Gao, Jianye Zhang, Xujie Liu, Hossein Heidari-Torkabadi, Phoebe L Stewart, William E Harte, Gregory P Tochtrop, Krzysztof Palczewski
ABSTRACT
Rhodopsin homeostasis is tightly coupled to rod photoreceptor cell survival and vision. Mutations resulting in the misfolding of rhodopsin can lead to autosomal dominant retinitis pigmentosa (adRP), a progressive retinal degeneration that currently is untreatable. Using a cell-based high-throughput screen (HTS) to identify small molecules that can stabilize the P23H-opsin mutant, which causes most cases of adRP, we identified a novel pharmacological chaperone of rod photoreceptor opsin, YC-001. As a non-retinoid molecule, YC-001 demonstrates micromolar potency and efficacy greater than 9-cis-retinal with lower cytotoxicity. YC-001 binds to bovine rod opsin with an EC50 similar to 9-cis-retinal. The chaperone activity of YC-001 is evidenced by its ability to rescue the transport of multiple rod opsin mutants in mammalian cells. YC-001 is also an inverse agonist that non-competitively antagonizes rod opsin signaling. Significantly, a single dose of YC-001 protects Abca4 -/- Rdh8 -/- mice from bright light-induced retinal degeneration, suggesting its broad therapeutic potential.
Sigma-Aldrich
4-(3-Butoxy-4-methoxybenzyl)imidazolidin-2-one, solid
Sigma-Aldrich
9-cis-Retinal, vitamin A analog