MilliporeSigma
  • SENP3 protects H9C2 cells from apoptosis triggered by H/R via STAT3 pathway.
SENP3 protects H9C2 cells from apoptosis triggered by H/R via STAT3 pathway.
European review for medical and pharmacological sciences (2018-05-18)
Y Zhang, L-M Zheng, C-X Wang, J-M Gu, S Xue
ABSTRACT
To investigate whether SENP3 protects H9C2 cells from apoptosis triggered by H/R through the signal transducer and activator of transcription 3 (STAT3) pathway. Male C57BL mice were cultured and mouse models of myocardial I/RI were established. At the same time, cardiomyoblast H9C2 cell line of rat embryo was cultured. Reactive oxygen species (ROS) level was detected during H/R using 2',7'-dichlorofluorescein diacetate (DCFH) kit. Apoptotic cells were checked by flow cytometry. The expressions of p-JAK2, JAK2, STAT3, p-STAT3, cleaved-caspase3 (c-caspase3), and Bcl/Bax were detected using Western blotting and reverse transcriptase-polymerase chain reaction (RT-PCR). We revealed that SENP3 rose in mice of I/R group and in H9C2 cells following H/R with an increase in p-STAT3. Furthermore, increased expression of SENP3 was found to be dependent on the generation of ROS, as the SENP3 accumulation was inhibited by antioxidant (NAC). Inhibition of SENP3 suppressed the p-STAT3 expression, but promoted cell apoptosis, c-caspase3 expression, and Bcl/Bax ratio. Besides, SENP3 overexpression alleviated the cell apoptosis, which was abrogated by AG490. SENP3 could protect H9C2 against H/R through enhancing JAK2/STAT3 pathway.