MilliporeSigma
  • Asymbiotic germination response to photoperiod and nutritional media in six populations of Calopogon tuberosus var. tuberosus (Orchidaceae): evidence for ecotypic differentiation.
Asymbiotic germination response to photoperiod and nutritional media in six populations of Calopogon tuberosus var. tuberosus (Orchidaceae): evidence for ecotypic differentiation.
Annals of botany (2008-09-02)
Philip J Kauth, Michael E Kane, Wagner A Vendrame, Carrie Reinhardt-Adams
ABSTRACT
Ecotypic differentiation has been explored in numerous plant species, but has been largely ignored in the Orchidaceae. Applying a specific germination protocol for widespread seed sources may be unreliable due to inherent physiological or genetic differences in localized populations. It is crucial to determine whether ecotypic differentiation exists for restoration and conservation programmes. Calopogon tuberosus var. tuberosus, a widespread terrestrial orchid of eastern North America, is a model species to explore ecotypic differences in germination requirements, as this species occupies diverse habitats spanning a wide geographical range. Mature seeds were collected from south Florida, north central Florida, three locations in South Carolina, and the upper Michigan peninsula. Effects of three photoperiods (8/16, 12/12, 16/8 h L/D) were examined on asymbiotic in vitro seed germination and seedling development of C. tuberosus. Germination and early development was monitored for 8 weeks, while advanced development was monitored for an additional 8 weeks. In an additional experiment, asymbiotic seed germination and development was monitored for 8 weeks on six culture media (BM-1 terrestrial orchid medium, Knudson C, Malmgrem, half-strength MS, P723, and Vacin and Went). A tetrazolium test for embryo viability was performed. Short days promoted the highest germination among Florida populations, but few differences among photoperiods in other seed sources existed. Different media had little effect on the germination of Michigan and Florida populations, but germination of South Carolina seeds was higher on media with higher calcium and magnesium. Tetrazolium testing confirmed that South Carolina seeds exhibited low viability while viability was higher in Florida seeds. Seed germination and corm formation was rapid in Michigan seeds across all treatments. Michigan seedlings allocated more biomass to corms compared with other seed sources. Rapid germination and corm formation may be a survival mechanism in response to a compressed growing season in northern populations. Ecotypic differentiation may be occurring based on seed germination and corm formation data.
Sigma-Aldrich
Knudson C Modified Orchid Medium, powder, suitable for plant cell culture