MilliporeSigma
  • PI(4,5)P2 controls plasma membrane PI4P and PS levels via ORP5/8 recruitment to ER-PM contact sites.
PI(4,5)P2 controls plasma membrane PI4P and PS levels via ORP5/8 recruitment to ER-PM contact sites.
The Journal of cell biology (2018-02-24)
Mira Sohn, Marek Korzeniowski, James P Zewe, Rachel C Wills, Gerald R V Hammond, Jana Humpolickova, Lukas Vrzal, Dominika Chalupska, Vaclav Veverka, Gregory D Fairn, Evzen Boura, Tamas Balla
ABSTRACT
Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2) is a critically important regulatory lipid of the plasma membrane (PM); however, little is known about how cells regulate PM PI(4,5)P2 levels. Here, we show that the phosphatidylinositol 4-phosphate (PI4P)/phosphatidylserine (PS) transfer activity of the endoplasmic reticulum (ER)-resident ORP5 and ORP8 is regulated by both PM PI4P and PI(4,5)P2 Dynamic control of ORP5/8 recruitment to the PM occurs through interactions with the N-terminal Pleckstrin homology domains and adjacent basic residues of ORP5/8 with both PI4P and PI(4,5)P2 Although ORP5 activity requires normal levels of these inositides, ORP8 is called on only when PI(4,5)P2 levels are increased. Regulation of the ORP5/8 attachment to the PM by both phosphoinositides provides a powerful means to determine the relative flux of PI4P toward the ER for PS transport and Sac1-mediated dephosphorylation and PIP 5-kinase-mediated conversion to PI(4,5)P2 Using this rheostat, cells can maintain PI(4,5)P2 levels by adjusting the availability of PI4P in the PM.
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
GSK-A1, ≥98% (HPLC)
Avanti
Click PI(4,5)P2-azido, Avanti Research - A Croda Brand 810340P, powder