MilliporeSigma
Improved nematode extraction from carrot disk culture.
Journal of nematology (1990-07-01)
D T Kaplan, E L Davis
ABSTRACT
Radopholus spp. were reared in carrot tissue culture via established procedures, with slight modification. Several plant tissue maceration enzymes and flotation media (salts and sucrose) were evaluated with regard to nematode toxicity and extraction efficiency. Best extraction of viable nematodes and eggs was attained when carrot tissue infested with Radopholus citrophilus or R. similis was macerated with a mixture of 0.50% driselase and 0.50% cellulysin, w/v each, with 2.5 ml of enzyme solution based for each gram of carrot tissue. Maceration slurries containing carrot tissue and nematodes were maintained in open flasks on a rotary shaker (175 rpm) at 26 C for 24 hours. Nematodes and eggs were extracted from resultant culture slurries by flotation with MgSO-7H0 (sp gr 1.1). A protocol is presented to extract large quantities of viable burrowing nematodes and their eggs from carrot disk cultures.
Sigma-Aldrich
Driselase Basidiomycetes sp., suitable for plant cell culture, BioReagent