MilliporeSigma
  • Rapid Changes in Plasma Membrane Protein Phosphorylation during Initiation of Cell Wall Digestion.
Rapid Changes in Plasma Membrane Protein Phosphorylation during Initiation of Cell Wall Digestion.
Plant physiology (1988-02-01)
D P Blowers, W F Boss, A J Trewavas
ABSTRACT
Plasma membrane vesicles from wild carrot cells grown in suspension culture were isolated by aqueous two-phase partitioning, and ATP-dependent phosphorylation was measured with [gamma-(32)P]ATP in the presence and absence of calcium. Treatment of the carrot cells with the cell wall digestion enzymes, driselase, in a sorbitol osmoticum for 1.5 min altered the protein phosphorylation pattern compared to that of cells treated with sorbitol alone. Driselase treatment resulted in decreased phosphorylation of a band of M(r) 80,000 which showed almost complete calcium dependence in the osmoticum treated cells; decreased phosphorylation of a band of M(r) 15,000 which showed little calcium activation, and appearance of a new band of calcium-dependent phosphorylation at M(r) 22,000. These effects appeared not to be due to nonspecific protease activity and neither in vivo nor in vitro exposure to driselase caused a significant loss of Coomassie blue-staining bands on the gels of the isolated plasma membranes. However, protein phosphorylation was decreased. Adding driselase to the in vitro reaction mixture caused a general decrease in the membrane protein phosphorylation either in the presence or absence of calcium which did not mimic the in vivo response. Cells labeled in vivo with inorganic (32)P also showed a response to the Driselase treatment. An enzymically active driselase preparation was required for the observed responses.
Sigma-Aldrich
Driselase Basidiomycetes sp., suitable for plant cell culture, BioReagent