MilliporeSigma
  • Dietary and Microbial Oxazoles Induce Intestinal Inflammation by Modulating Aryl Hydrocarbon Receptor Responses.
Dietary and Microbial Oxazoles Induce Intestinal Inflammation by Modulating Aryl Hydrocarbon Receptor Responses.
Cell (2018-05-19)
Shankar S Iyer, Thomas Gensollen, Amit Gandhi, Sungwhan F Oh, Joana F Neves, Frederic Collin, Richard Lavin, Carme Serra, Jonathan Glickman, Punyanganie S A de Silva, R Balfour Sartor, Gurdyal Besra, Russell Hauser, Anthony Maxwell, Amadeu Llebaria, Richard S Blumberg
ABSTRACT
Genome-wide association studies have identified risk loci associated with the development of inflammatory bowel disease, while epidemiological studies have emphasized that pathogenesis likely involves host interactions with environmental elements whose source and structure need to be defined. Here, we identify a class of compounds derived from dietary, microbial, and industrial sources that are characterized by the presence of a five-membered oxazole ring and induce CD1d-dependent intestinal inflammation. We observe that minimal oxazole structures modulate natural killer T cell-dependent inflammation by regulating lipid antigen presentation by CD1d on intestinal epithelial cells (IECs). CD1d-restricted production of interleukin 10 by IECs is limited through activity of the aryl hydrocarbon receptor (AhR) pathway in response to oxazole induction of tryptophan metabolites. As such, the depletion of the AhR in the intestinal epithelium abrogates oxazole-induced inflammation. In summary, we identify environmentally derived oxazoles as triggers of CD1d-dependent intestinal inflammatory responses that occur via activation of the AhR in the intestinal epithelium.
Sigma-Aldrich
MISSION® esiRNA, targeting human AHR, RP11-507K12.1
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
2,4,5-Trimethyl-2,5-dihydro-1,3-oxazole, AldrichCPR
Supelco
Vinclozolin, PESTANAL®, analytical standard
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
2-Methyl-1-pyrroline, 95%
Sigma-Aldrich
(2S,3S,4R)-1-O-(α-D-Galactosyl)-N-hexacosanoyl-2-amino-1,3,4-octadecanetriol, ≥95% (TLC)