MilliporeSigma
  • Improved LC-MS/MS method for the quantification of hepcidin-25 in clinical samples.
Improved LC-MS/MS method for the quantification of hepcidin-25 in clinical samples.
Analytical and bioanalytical chemistry (2018-04-19)
Ioana M Abbas, Holger Hoffmann, María Montes-Bayón, Michael G Weller
ABSTRACT
Mass spectrometry-based methods play a crucial role in the quantification of the main iron metabolism regulator hepcidin by singling out the bioactive 25-residue peptide from the other naturally occurring N-truncated isoforms (hepcidin-20, -22, -24), which seem to be inactive in iron homeostasis. However, several difficulties arise in the MS analysis of hepcidin due to the "sticky" character of the peptide and the lack of suitable standards. Here, we propose the use of amino- and fluoro-silanized autosampler vials to reduce hepcidin interaction to laboratory glassware surfaces after testing several types of vials for the preparation of stock solutions and serum samples for isotope dilution liquid chromatography-tandem mass spectrometry (ID-LC-MS/MS). Furthermore, we have investigated two sample preparation strategies and two chromatographic separation conditions with the aim of developing a LC-MS/MS method for the sensitive and reliable quantification of hepcidin-25 in serum samples. A chromatographic separation based on usual acidic mobile phases was compared with a novel approach involving the separation of hepcidin-25 with solvents at high pH containing 0.1% of ammonia. Both methods were applied to clinical samples in an intra-laboratory comparison of two LC-MS/MS methods using the same hepcidin-25 calibrators with good correlation of the results. Finally, we recommend a LC-MS/MS-based quantification method with a dynamic range of 0.5-40 μg/L for the assessment of hepcidin-25 in human serum that uses TFA-based mobile phases and silanized glass vials. Graphical abstract Structure of hepcidin-25 (Protein Data Bank, PDB ID 2KEF).
Product Description
Supelco
Certified Vial Kit, Low Adsorption (LA) QsertVial, volume 0.3 mL, QsertVial, clear glass vial, natural PTFE/silicone septa, thread for 9 mm
Sigma-Aldrich
Sheep Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)