MilliporeSigma
Luminol chemiluminescence under interaction with heteropoly acids.
Talanta (2009-03-10)
Oleg Zui, Hiroki Takahashi, Toshitaka Hori, Teruo Hinoue
ABSTRACT
Interaction of luminol with phosphomolybdic, phosphovanadomolybdic and silicomolybdic acids was studied by examination of chemiluminescence spectra, measurement of ESR spectra, investigation of reaction order, and elucidation of inhibition effects. A scheme of the reaction mechanism is proposed.
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent