MilliporeSigma
  • Thermally stable hybrid polyarylidene(azomethine-ether)s polymers (PAAP): an ultrasensitive arsenic(III) sensor approach.
Thermally stable hybrid polyarylidene(azomethine-ether)s polymers (PAAP): an ultrasensitive arsenic(III) sensor approach.
Designed monomers and polymers (2018-05-31)
Mohammed M Rahman, Mahmoud A Hussein, Kamal I Aly, Abdullah M Asiri
ABSTRACT
A new category of thermally stable hybrid polyarylidene(azomethine-ether)s and copolyarylidene(azomethine-ether)s (PAAP) based on diarylidenecycloalkanones has been synthesized using solution polycondensation method. For potential cationic sensor development, a thin layer of PAAP onto a flat glassy carbon electrode (GCE, surface area: 0.0316 cm2) was prepared with conducting nafion (5%) coating agent to fabricate a sensitive and selective arsenic (III) [As3+] ion in short response time in neutral buffer system. The fabricated cationic sensor was measured the analytical performances such as higher sensitivity, linear dynamic range, detection limit, reproducibility, and long-term stability towards As3+ ions. The sensitivity and detection limit were calculated as 2.714 μAμM-1cm-2 and 6.8 ± 0.1 nM (SNR of 3; 3N/S) respectively from the calibration curve. This novel approach can be initiated a well-organized route of an efficient development of heavy selective arsenic sensor for hazardous pollutants in biological, environmental, and health care fields. Real sample analysis for various environmental sample was performed with PAAP-modified-GCE.
Sigma-Aldrich
4-Chlorobenzaldehyde, 97%