MilliporeSigma
  • Quantifying absolute addressability in DNA origami with molecular resolution.
Quantifying absolute addressability in DNA origami with molecular resolution.
Nature communications (2018-04-25)
Maximilian T Strauss, Florian Schueder, Daniel Haas, Philipp C Nickels, Ralf Jungmann
ABSTRACT
Self-assembled DNA nanostructures feature an unprecedented addressability with sub-nanometer precision and accuracy. This addressability relies on the ability to attach functional entities to single DNA strands in these structures. The efficiency of this attachment depends on two factors: incorporation of the strand of interest and accessibility of this strand for downstream modification. Here we use DNA-PAINT super-resolution microscopy to quantify both incorporation and accessibility of all individual strands in DNA origami with molecular resolution. We find that strand incorporation strongly correlates with the position in the structure, ranging from a minimum of 48% on the edges to a maximum of 95% in the center. Our method offers a direct feedback for the rational refinement of the design and assembly process of DNA nanostructures and provides a long sought-after quantitative explanation for efficiencies of DNA-based nanomachines.
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder