MilliporeSigma
Nickel-catalyzed Negishi alkylations of styrenyl aziridines.
Journal of the American Chemical Society (2012-03-15)
Chung-Yang Huang, Abigail G Doyle
ABSTRACT
A nickel-catalyzed cross-coupling reaction between N-sulfonyl aziridines and organozinc reagents is reported. The catalytic system comprises an inexpensive and air-stable Ni(II) source and dimethyl fumarate as ligand. Regioselective synthesis of β-substituted amines is possible under mild and functional-group-tolerant conditions. The stereoselectivity of the reaction is consistent with a stereoconvergent mechanism wherein the sulfonamide directs C-C bond formation.
Sigma-Aldrich
Dimethyl fumarate, 97%