MilliporeSigma
  • In vitro cytotoxicity assessment of nanodiamond particles and their osteogenic potential.
In vitro cytotoxicity assessment of nanodiamond particles and their osteogenic potential.
Journal of biomedical materials research. Part A (2018-02-17)
Mohamed Ibrahim, Ying Xue, Melanie Ostermann, Alexander Sauter, Doris Steinmueller-Nethl, Sarah Schweeberg, Anke Krueger, Mihaela R Cimpan, Kamal Mustafa
ABSTRACT
Scaffolds functionalized with nanodiamond particles (nDP) hold great promise with regard to bone tissue formation in animal models. Degradation of the scaffolds over time may leave nDP within the tissues, raising concerns about possible long-term unwanted effects. Human SaOS-2 osteoblast-like cells and U937 monoblastoid cells were exposed to five different concentrations (0.002-2 mg/L) of nDP (size range: 2.36-4.42 nm) for 24 h. Cell viability was assessed by impedance-based methods. The differential expression of stress and toxicity-related genes was evaluated by polymerase chain reaction (PCR) super-array, while the expression of selected inflammatory and cell death markers was determined by reverse transcriptase quantitative polymerase chain reaction (RT-qPCR). Furthermore, the expression of osteogenic genes by SaOS-2 cells, alkaline phosphatase activity and the extracellular calcium nodule deposition in response to nDP were determined in vitro. Cells responded differently to higher nDP concentrations (≥0.02 mg/L), that is, no loss of viability for SaOS-2 cells and significantly reduced viability for U937 cells. Gene expression showed significant upregulation of several cell death and inflammatory markers, among other toxicity reporter genes, indicating inflammatory and cytotoxic responses in U937 cells. Nanodiamond particles improved the osteogenicity of osteoblast-like cells with no evident cytotoxicity. However, concentration-dependent cytotoxic and inflammatory responses were seen in the U937 cells, negatively affecting osteogenicity in co-cultures. © 2018 Wiley Periodicals, Inc. J Biomed Mater Res Part A: 106A: 1697-1707, 2018.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Alizarin, Dye content 97 %