MilliporeSigma
  • Contribution of mGluR5 to pathophysiology in a mouse model of human chromosome 16p11.2 microdeletion.
Contribution of mGluR5 to pathophysiology in a mouse model of human chromosome 16p11.2 microdeletion.
Nature neuroscience (2015-01-13)
Di Tian, Laura J Stoppel, Arnold J Heynen, Lothar Lindemann, Georg Jaeschke, Alea A Mills, Mark F Bear
ABSTRACT
Human chromosome 16p11.2 microdeletion is the most common gene copy number variation in autism, but the synaptic pathophysiology caused by this mutation is largely unknown. Using a mouse with the same genetic deficiency, we found that metabotropic glutamate receptor 5 (mGluR5)-dependent synaptic plasticity and protein synthesis was altered in the hippocampus and that hippocampus-dependent memory was impaired. Notably, chronic treatment with a negative allosteric modulator of mGluR5 reversed the cognitive deficit.
Sigma-Aldrich
CTEP, ≥98% (HPLC)