MilliporeSigma
  • High Efficacy of Finafloxacin on Helicobacter pylori Isolates at pH 5.0 Compared with That of Other Fluoroquinolones.
High Efficacy of Finafloxacin on Helicobacter pylori Isolates at pH 5.0 Compared with That of Other Fluoroquinolones.
Antimicrobial agents and chemotherapy (2015-09-30)
Jung Won Lee, Nayoung Kim, Ryoung Hee Nam, Jung Mogg Kim, Jong Youn Park, Sun Min Lee, Joo Sung Kim, Dong Ho Lee, Hyun Chae Jung
ABSTRACT
Finafloxacin is a novel fluoroquinolone with improved antimicrobial efficacy, especially in an acidic environment. The efficacy of finafloxacin for the inhibition of Helicobacter pylori infection was compared with the efficacies of levofloxacin and moxifloxacin at neutral and acidic pH. The impacts of gyrA point mutation on the efficacy of those three fluoroquinolones were also investigated. A total of 128 clinical H. pylori strains were utilized. MICs of levofloxacin, moxifloxacin, and finafloxacin were determined at pH 5.0 and pH 7.0 by the agar dilution method. The impact of gyrA point mutations that are responsible for fluoroquinolone resistance was analyzed; the results showed 50 strains with an Asn-87 point mutation, 48 strains with an Asp-91 point mutation, and the remaining 30 strains with no gyrA mutations. The use of finafloxacin led to MIC values at pH 5.0 that were lower than the values seen at pH 7.0 for 112 strains (112/128, 87.5%), and this proportion was higher than that seen with moxifloxacin (21/128, 16.4%, P < 0.001). Finafloxacin also demonstrated a rate of susceptibility (MIC, <1 μg/ml) (37.5%, 48/128) at pH 5.0 that was higher than that seen with moxifloxacin (2.3%, 3/128) (P < 0.001). The trends were similar regardless of which of the Asn-87, Asp-91, and A2143 point mutations were present. In conclusion, the superior antimicrobial efficacy of finafloxacin against H. pylori in an acidic environment suggests the possible use of finafloxacin for treatment of H. pylori infection, as has been proposed by its developer, Merlion Pharma.
Sigma-Aldrich
Finafloxacin, ≥95% (HPLC)