MilliporeSigma
  • Improvement of postoperative cognitive dysfunction and attention network function of patients with ischemic cerebrovascular disease via dexmedetomidine.
Improvement of postoperative cognitive dysfunction and attention network function of patients with ischemic cerebrovascular disease via dexmedetomidine.
Experimental and therapeutic medicine (2018-03-31)
Jingchao Zhang, Guoliang Wang, Fangxiang Zhang, Qian Zhao
ABSTRACT
The protective effect of dexmedetomidine on cognitive dysfunction and decreased attention network function of patients with ischemic cerebrovascular disease after stenting was investigated. Fifty-eight patients with ischemic cerebrovascular disease undergoing stenting in Guizhou Provincial People's Hospital were selected and randomly divided into control group (n=29) and dexmedetomidine group (n=29). The dexmedetomidine group was treated with dexmedetomidine before induced anesthesia, while the control group was given the same dose of normal saline; and the normal volunteers of the same age were selected as the normal group (n=29). At 3 days after operation, the levels of serum S100B and nerve growth factor (NGF) in each group were detected using the enzyme-linked immunosorbent assay, and the level of brain-derived neurotrophic factor (BDNF) was detected via western blotting. Montreal cognitive assessment (MoCA) and attention network test (ANT) were performed. Moreover, the cognitive function and attention network function, and the effects of dexmedetomidine on cognitive function and attention network function were evaluated. The concentrations of serum S100B and NGF in dexmedetomidine group was lower than those in control group (P<0.01). The results of western blotting showed that the levels of serum BDNF in control group and dexmedetomidine group were significantly lower than that in normal group (P<0.01), and it was higher in dexmedetomidine group than that in control group (P<0.01). Besides, both MoCA and ANT results revealed that the visual space and executive function scores, attention scores, delayed memory scores, targeted network efficiency and executive control network efficiency in dexmedetomidine group were obviously higher than those in control group (P<0.01). The cognitive function and attention network function of patients with ischemic cerebrovascular disease have a certain degree of damage, and the preoperative administration of dexmedetomidine can effectively improve the patient's cognitive dysfunction and attention network function after operation.
Sigma-Aldrich
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-BDNF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution