MilliporeSigma
  • MCT4 as a potential therapeutic target for metastatic gastric cancer with peritoneal carcinomatosis.
MCT4 as a potential therapeutic target for metastatic gastric cancer with peritoneal carcinomatosis.
Oncotarget (2016-05-26)
Ji Yun Lee, InKyoung Lee, Won Jin Chang, Su Min Ahn, Sung Hee Lim, Hae Su Kim, Kwai Han Yoo, Ki Sun Jung, Haa-Na Song, Jin Hyun Cho, Sun Young Kim, Kyoung-Mee Kim, Soojin Lee, Seung Tae Kim, Se Hoon Park, Jeeyun Lee, Joon Oh Park, Young Suk Park, Ho Yeong Lim, Won Ki Kang
ABSTRACT
Monocarboxylate transporters (MCTs) play a major role in up-regulation of glycolysis and adaptation to acidosis. However, the role of MCTs in gastric cancer (GC) is not fully understood. We investigated the potential utilization of a new cancer therapy for GC. We characterized the expression patterns of the MCT isoforms 1, 2, and 4 and investigated the role of MCT in GC through in vitro and in vivo tests using siRNA targeting MCTs. In GC cell lines, MCT1, 2, and 4 were up-regulated with different expression levels; MCT1 and MCT4 were more widely expressed in GC cell lines compared with MCT2. Inhibition of MCTs by siRNA or AR-C155858 reduced cell viability and lactate uptake in GC cell lines. The effect of inhibition of MCTs on tumor growth was also confirmed in xenograft models. Furthermore, MCT inhibition in GC cells increased the sensitivity of cells to radiotherapy or chemotherapy. Compared with normal gastric tissue, no significant alterations of expression levels in tumors were identified for MCT1 and MCT2, whereas a significant increase in MCT4 expression was observed. Most importantly, MCT4 was highly overexpressed in malignant cells of acsites and its silencing resulted in reduced tumor cell proliferation and lactate uptake in malignant ascites. Our study suggests that MCT4 is a clinically relevant target in GC with peritoneal carcinomatosis.
Sigma-Aldrich
Anti-SLC16A7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.