MilliporeSigma
  • Involvement of B Cells, Immunoglobulins, and Syk in the Pathogenesis of Abdominal Aortic Aneurysm.
Involvement of B Cells, Immunoglobulins, and Syk in the Pathogenesis of Abdominal Aortic Aneurysm.
Journal of the American Heart Association (2018-03-17)
Aya Furusho, Hiroki Aoki, Satoko Ohno-Urabe, Michihide Nishihara, Saki Hirakata, Norifumi Nishida, Sohei Ito, Makiko Hayashi, Tsutomu Imaizumi, Shinichi Hiromatsu, Hidetoshi Akashi, Hiroyuki Tanaka, Yoshihiro Fukumoto
ABSTRACT
Abdominal aortic aneurysm (AAA) is a potentially life-threatening disease that is common in older individuals. Currently, therapeutic options are limited to surgical interventions. Although it has long been known that AAA tissue is enriched in B cells and immunoglobulins, their involvement in AAA pathogenesis remains controversial. We investigated the role of B cells and immunoglobulins in a murine model of AAA, induced with a periaortic application of CaCl2, and in human AAA. Both human and mouse AAA tissue showed B-cell infiltration. Mouse AAA tissue showed deposition of IgG and activation of Syk, a key molecule in B-cell activation and immunoglobulin function, which were localized to infiltrating cells including B cells and macrophages. B-cell-deficient muMT mice showed suppression of AAA development that was associated with reduced activation of Syk and less expression of matrix metalloproteinase-9. Administration of exogenous immunoglobulins restored the blunted Syk activation and AAA development in muMT mice. Additionally, exogenous immunoglobulins induced interleukin-6 and metalloproteinase-9 secretions in human AAA tissue cultures. Furthermore, administration of R788, a specific Syk inhibitor, suppressed AAA expansion, reduced inflammatory response, and reduced immunoglobulin deposition in AAA tissue. From these results, we concluded that B cells and immunoglobulins participated in AAA pathogenesis by promoting inflammatory and tissue-destructive activities. Finally, we identified Syk as a potential therapeutic target.
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture