MilliporeSigma
  • Mutations in L-type amino acid transporter-2 support SLC7A8 as a novel gene involved in age-related hearing loss.
Mutations in L-type amino acid transporter-2 support SLC7A8 as a novel gene involved in age-related hearing loss.
eLife (2018-01-23)
Meritxell Espino Guarch, Mariona Font-Llitjós, Silvia Murillo-Cuesta, Ekaitz Errasti-Murugarren, Adelaida M Celaya, Giorgia Girotto, Dragana Vuckovic, Massimo Mezzavilla, Clara Vilches, Susanna Bodoy, Ignasi Sahún, Laura González, Esther Prat, Antonio Zorzano, Mara Dierssen, Isabel Varela-Nieto, Paolo Gasparini, Manuel Palacín, Virginia Nunes
ABSTRACT
Age-related hearing loss (ARHL) is the most common sensory deficit in the elderly. The disease has a multifactorial etiology with both environmental and genetic factors involved being largely unknown. SLC7A8/SLC3A2 heterodimer is a neutral amino acid exchanger. Here, we demonstrated that SLC7A8 is expressed in the mouse inner ear and that its ablation resulted in ARHL, due to the damage of different cochlear structures. These findings make SLC7A8 transporter a strong candidate for ARHL in humans. Thus, a screening of a cohort of ARHL patients and controls was carried out revealing several variants in SLC7A8, whose role was further investigated by in vitro functional studies. Significant decreases in SLC7A8 transport activity was detected for patient's variants (p.Val302Ile, p.Arg418His, p.Thr402Met and p.Val460Glu) further supporting a causative role for SLC7A8 in ARHL. Moreover, our preliminary data suggest that a relevant proportion of ARHL cases could be explained by SLC7A8 mutations.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra