MilliporeSigma
  • The glutathione dependence of inorganic sulfate formation from L- or D-cysteine in isolated rat hepatocytes.
The glutathione dependence of inorganic sulfate formation from L- or D-cysteine in isolated rat hepatocytes.
Chemico-biological interactions (1998-06-03)
J Huang, S Khan, P J O'Brien
ABSTRACT
The GSH dependence of the metabolic pathways involved in the conversion of cysteine to sulfate in intact cells has been investigated. It was found that hepatocyte-catalysed sulfate formation from added L-cysteine did not occur if hepatocyte GSH was depleted beforehand, but was restored when GSH levels recovered. Furthermore, sulfate formation did not recover in GSH-depleted hepatocytes if GSH synthesis was prevented with buthionine sulfoximine. Thiosulfate formation was, however, markedly enhanced in GSH-depleted hepatocytes. These results suggest that thiosulfate is an intermediate in the formation of inorganic sulfate from L-cysteine and that GSH was required for the conversion of thiosulfate to inorganic sulfate. Much less sulfate was formed if the cysteine was replaced with cysteinesulfinate. Furthermore, sulfate formation from L-cysteine was markedly inhibited by the addition of the transaminase inhibitor DL-cycloserine or the gamma-cystathionase inhibitor DL-propargylglycine. The major routes of sulfate formation from L-cysteine therefore seems to involve pathways that do not involve L-cysteinesulfinate. Similar amounts of sulfate were formed from D-cysteine as L-cysteine. Thiosulfate instead of sulfate was also formed in GSH-depleted hepatocytes. However, sulfate formation from D-cysteine differed from L-cysteine in that it was inhibited by the D-aminoacid oxidase inhibitor sodium benzoate and was not affected by transaminase or gamma-cystathionase inhibitors. These results suggest that thiosulfate is an intermediate in sulfate formation from D-cysteine and involves the oxidation of D-cysteine by D-amino acid oxidase to form beta-mercaptopyruvate.