MilliporeSigma
  • Facilitated transport of two model steroids by esters and amides of clofibric acid.
Facilitated transport of two model steroids by esters and amides of clofibric acid.
Journal of pharmaceutical sciences (1993-02-01)
B B Michniak, J M Chapman, K L Seyda
ABSTRACT
A series of novel dermal penetration enhancers, esters and amides of clofibric acid, was synthesized. The permeation parameters and skin retention of two steroids (hydrocortisone-21-acetate and betamethasone-17-valerate) in propylene glycol were studied with athymic nude mouse skin by in vitro diffusion cell techniques in the presence of the novel enhancer compounds. Isopropyl myristate, dimethyl lauramide, and 1-dodecylazacycloheptan-2-one (laurocapram, Azone) were used as control enhancers. The most satisfactory enhancement of both the ester and amide series was observed with clofibric acid octyl amide; coadministration increased skin retention of hydrocortisone acetate after 24 h by 3.5-fold and that of betamethasone valerate by 2.9-fold. Diffusion cell receptor concentrations increased 51.6- and 10.3-fold, respectively, during the same time period. However, the enhancer compound in this case was applied to the skin 1 h prior to each of the steroids. The amide analogues were more effective than the equivalent ester compounds of the same carbon chain length. The best enhancer compounds (2c, 3d, 3e, and 3f) were nonirritating to athymic mouse skin in vivo.