MilliporeSigma
  • Chondrocyte-specific Smad4 gene conditional knockout results in hearing loss and inner ear malformation in mice.
Chondrocyte-specific Smad4 gene conditional knockout results in hearing loss and inner ear malformation in mice.
Developmental dynamics : an official publication of the American Association of Anatomists (2009-07-08)
Shi-ming Yang, Zhao-hui Hou, Guan Yang, Ji-shuai Zhang, Yin-yan Hu, Jian-he Sun, Wei-wei Guo, David z z He, Dong-yi Han, Wie-yen Young, Xiao Yang
ABSTRACT
Smad4 is the central intracellular mediator of transforming growth factor-beta (TGF-beta) signaling, which plays crucial roles in tissue regeneration, cell differentiation, embryonic development, and regulation of the immune system. Conventional Smad4 gene knockout results in embryonic lethality, precluding its use in studies of the role of Smad4 in inner ear development. We used chondrocyte-specific Smad4 knockout mice (Smad4Co/Co) to investigate the function of Smad4 in inner ear development. Smad4Co/Co mice were characterized by a smaller cochlear volume, bone malformation, and abnormalities of the osseous spiral lamina and basilar membrane. The development of the hair cells was also abnormal, as evidenced by the disorganized stereocilia and reduced density of the neuronal processes beneath the hair cells. Auditory function tests revealed the homozygous Smad4Co/Co mice suffered from severe sensorineural hearing loss. Our results suggest that Smad4 is required for inner ear development and normal auditory function in mammals.
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Anti-Myosin VI (KA-15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution