MilliporeSigma
  • Cardiac, Metabolic and Molecular Profiles of Sedentary Rats in the Initial Moment of Obesity.
Cardiac, Metabolic and Molecular Profiles of Sedentary Rats in the Initial Moment of Obesity.
Arquivos brasileiros de cardiologia (2017-10-27)
Bruno Barcellos Jacobsen, Ana Paula Lima Leopoldo, Jóctan Pimentel Cordeiro, Dijon Henrique Salomé de Campos, André Ferreira do Nascimento, Mário Mateus Sugizaki, Antônio Carlos Cicogna, Carlos Roberto Padovani, André Soares Leopoldo
ABSTRACT
Different types of high-fat and/or high-energy diets have been used to induce obesity in rodents. However, few studies have reported on the effects observed at the initial stage of obesity induced by high-fat feeding on cardiac functional and structural remodelling. To characterize the initial moment of obesity and investigate both metabolic and cardiac parameters. In addition, the role of Ca2+ handling in short-term exposure to obesity was verified. Thirty-day-old male Wistar rats were randomized into two groups (n = 19 each): control (C; standard diet) and high-fat diet (HF, unsaturated high-fat diet). The initial moment of obesity was defined by weekly measurement of body weight (BW) complemented by adiposity index (AI). Cardiac remodelling was assessed by morphological, histological, echocardiographic and papillary muscle analysis. Ca2+ handling proteins were determined by Western Blot. The initial moment of obesity occurred at the 3rd week. Compared with C rats, the HF rats had higher final BW (4%), body fat (20%), AI (14.5%), insulin levels (39.7%), leptin (62.4%) and low-density lipoprotein cholesterol (15.5%) but did not exhibit alterations in systolic blood pressure. Echocardiographic evaluation did not show alterations in cardiac parameters. In the HF group, muscles were observed to increase their +dT/dt (C: 52.6 ± 9.0 g/mm2/s and HF: 68.0 ± 17.0 g/mm2/s; p < 0.05). In addition, there was no changes in the cardiac expression of Ca2+ handling proteins. The initial moment of obesity promotes alterations to hormonal and lipid profiles without cardiac damage or changes in Ca2+ handling.
Sigma-Aldrich
Anti-Calcium Channel (Cardiac α1C Subunit) (L-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Pricing and availability is not currently available.