MilliporeSigma
  • Specific inhibition of phorbol ester and DiC8-induced histamine release from human basophils by the protein kinase C inhibitors, H-7 and H-9.
Specific inhibition of phorbol ester and DiC8-induced histamine release from human basophils by the protein kinase C inhibitors, H-7 and H-9.
Biochemical and biophysical research communications (1987-04-29)
D O Thueson, J A Kennedy, C D Wright, M C Conroy
ABSTRACT
The release of histamine and other inflammatory mediators from human basophils is triggered by numerous stimuli, including chemical, physical and receptor-mediated activators. Several mechanisms of cell activation including protein kinase C activation have been proposed to operate in these cells. We used phorbol ester and DiC8 to induce histamine release from human basophils and the protein kinase C inhibitors H-7 and H-9 to inhibit this release. Both DiC8 and TPA induced histamine release were inhibited by H-7 (ID 50 = 37 mcM) and H-9 (IC 50 = 20 mcM). However, anti-IgE, fmlp and A23187-induced histamine release were unaffected. In contrast, the calmodulin antagonists W-7 and perphenazine effectively inhibited histamine release by all five stimuli. Therefore, different biochemical pathways appear to be critical for basophil activation depending on the nature of the stimulus used.