MilliporeSigma
  • Timely regulated sorting from early to late endosomes is required to maintain cerebellar long-term depression.
Timely regulated sorting from early to late endosomes is required to maintain cerebellar long-term depression.
Nature communications (2017-09-03)
Taegon Kim, Yukio Yamamoto, Keiko Tanaka-Yamamoto
ABSTRACT
An important feature of long-term synaptic plasticity is the prolonged maintenance of plastic changes in synaptic transmission. The trafficking of AMPA-type glutamate receptors (AMPARs) is involved in the expression of many forms of synaptic plasticity, yet the subsequent events accomplishing the maintenance of plastic changes in synaptic AMPAR numbers are not fully understood. Here, we find that maintenance of cerebellar long-term depression results from a reduction in the number of AMPARs residing within endocytic recycling pathways. We then develop a genetically encoded, photosensitive inhibitor of late endosome sorting and use this to discover that initial maintenance of long-term depression relies on timely regulated late endosome sorting, which exhibits a threshold as well as switch-like behavior. Thus, our results indicate that recycling AMPAR numbers are reduced by a switching machinery of transient late endosome sorting, and that this process enables the transition from basal synaptic transmission to long-term depression maintenance.Long term depression (LTD) of the cerebellum is known to be mediated by postsynaptic trafficking of glutamate receptor AMPAR. Here, Kim and colleagues show that early- to late-endosomal sorting of AMPAR represents the switch from expression to maintenance phase of cerebellar LTD.
Sigma-Aldrich
Anti-EEA1 antibody, Mouse monoclonal, clone EEA1-N19, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rab7 antibody, Mouse monoclonal, ~2 mg/mL, clone Rab7-117, purified from hybridoma cell culture
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)