MilliporeSigma
  • microRNAs and DICER1 are regulated by 1,25-dihydroxyvitamin D in prostate stroma.
microRNAs and DICER1 are regulated by 1,25-dihydroxyvitamin D in prostate stroma.
The Journal of steroid biochemistry and molecular biology (2017-01-17)
Shweta Dambal, Angeline A Giangreco, Andres M Acosta, Andrew Fairchild, Zachary Richards, Ryan Deaton, Dennis Wagner, Reinhold Vieth, Peter H Gann, Andre Kajdacsy-Balla, Theodorus Van der Kwast, Larisa Nonn
ABSTRACT
Vitamin D deficiency increases the risk of lethal prostate adenocarcinomas (PCa) and the majority of older men are deficient. Although PCa arises from the epithelium, the surrounding stroma has hormonal regulatory control over the epithelium and contributes to carcinogenesis. Herein, we describe regulation of microRNAs (miRs) by the active hormone dihydroxyvitamin D (1,25(OH)2D) in human prostate stroma. 1,25(OH)2D binds the vitamin D receptor (VDR) transcription factor to regulate gene expression, including miRs, which have emerged as potent regulators of protein expression. 1,25(OH)2D-regulated miRs were identified by profiling in primary human prostatic stromal cells (PrS) and three miRs, miR-126-3p, miR 154-5p and miR-21-5p were subsequently validated in laser-capture micro-dissected prostate stromal tissue from a vitamin D3 clinical trial (N=45). Regulation of these miRs by 1,25(OH)2D was VDR-dependent. Network analysis of known and putative mRNA targets of these miRs was enriched with cancer and inflammation pathways, consistent with known roles of stroma and of vitamin D in carcinogenesis. Expression of the miR processing ribonuclease, DICER1, positively correlated with vitamin D metabolite levels in the clinical trial specimens. High epithelial/stromal ratios of DICER1 were significantly associated biochemical recurrence (OR 3.1, p=0.03) in a tissue microarray of 170 matched PCa patients. In summary, these results underscore the role of the prostate stroma in regulating responses to the hormone 1,25(OH)2D and identified miRs and DICER1 as being regulated in human prostate stroma. Regulation of stromal DICER1 by 1,25(OH)2D may also have clinical relevance in protection against aggressive PCa.
Sigma-Aldrich
Anti-DICER1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution