MilliporeSigma
  • Aminoglutethimide medical adrenalectomy for advanced prostatic carcinoma.
Aminoglutethimide medical adrenalectomy for advanced prostatic carcinoma.
The Journal of urology (1976-02-01)
E J Sanford, J R Drago, T J Rohner, R Santen, A Lipton
PMID765499
ABSTRACT
Complete adrenal suppression with aminoglutethimide has been accomplished in 7 patients with progressive stage D carcinoma of the prostate who had become refractory to orchiectomy and the administration of exogenous estrogens. A favorable response was noted in 3 patients. These preliminary results indicate that this agent may be as useful as surgical adrenalectomy in the treatment of progressive prostatic carcinoma. A brief discussion of the pharmacology of this agent, its mode of administration and the side effects are presented.
Sigma-Aldrich
DL-Aminoglutethimide