MilliporeSigma
  • Methamphetamine and Ovarian Steroid Responsive Cells in the Posteriodorsal Medial Amygdala are Required for Methamphetamine-enhanced Proceptive Behaviors.
Methamphetamine and Ovarian Steroid Responsive Cells in the Posteriodorsal Medial Amygdala are Required for Methamphetamine-enhanced Proceptive Behaviors.
Scientific reports (2017-01-04)
Katrina M Williams, Jessica A Mong
ABSTRACT
Methamphetamine (Meth) is a psychomotor stimulant strongly associated with increases in sexual drive and impulse in both men and women. These changes in sexual motivation have a greater impact on women due to their likelihood of facing the greater burden of unplanned pregnancies, as well as increased risk for psychiatric co-morbidities such as depression. We have previously established a rodent model of Meth-induced increases in sexual motivation. Using this model, we have identified the posteriodorsal medial amygdala (MePD) via excitotoxic lesion studies as a necessary nucleus in Meth-facilitated female sexual motivation. While lesion studies give us insight into key nuclei that may be targets of Meth action, such an approach does not give insight into the identity of the specific MePD neurons or neural circuitry involved in Meth-induced increases in proceptive behaviors. Using the DAUN02 inactivation method, a recently established technique for removing behaviorally relevant cell populations, we present evidence that the ovarian steroid/Meth responsive cells in the MePD are necessary for Meth-induced facilitation of proceptive behaviors. These findings form the basis for future work that will allow for the classification of neuronal subtypes involved in the MePD's modulation of proceptive behavior as well as a stronger understanding of the neurocircuitry of female sexual motivation.
Sigma-Aldrich
Monoclonal Anti-β-Galactosidase antibody produced in mouse, clone GAL-13, ascites fluid