MilliporeSigma
  • Role of monocyte chemoattractant protein-1 in liver fibrosis with transient myeloproliferative disorder in down syndrome.
Role of monocyte chemoattractant protein-1 in liver fibrosis with transient myeloproliferative disorder in down syndrome.
Hepatology communications (2018-03-07)
Kenichiro Kobayashi, Takako Yoshioka, Jun Miyauchi, Atsuko Nakazawa, Nobutaka Kiyokawa, Toshiro Maihara, Ikuya Usami
ABSTRACT
Liver fibrosis is a common complication associated with transient myeloproliferative disorder (TMD) in Down syndrome (DS). The exact molecular pathogenesis that regulates disease progression is largely unknown. We recently found serum and/or urinary monocyte chemoattractant protein-1 (MCP-1) as a novel biomarker of liver fibrosis. This study was an in vitro analysis to investigate the fibrogenic activity of MCP-1 using the collagen-producing LX-2 human hepatic stellate cell line. We also examined the fibrogenic activity of serum from a male neonate with DS in whom late-onset liver fibrosis developed even after the resolution of TMD. MCP-1 stimulated both cell growth and collagen synthesis of LX-2 in a dose-dependent manner. Patient serum obtained during the active disease phase significantly up-regulated fibrogenic activity, which was suppressed in the presence of MCP-1-blocking antibody. Transient transforming growth factor beta 1 stimulation primed LX-2 to induce prolonged hypersecretion of MCP-1 in the culture supernatant and in collagen synthesis, which was suppressed with MCP-1 blocking antibody as well. Conclusion: MCP-1 accounts for the prolonged activation of collagen-producing hepatic stellate cells in both a paracrine and autocrine manner, thereby promoting liver fibrosis. Anti-cytokine therapy targeting the fibrogenic cytokines of MCP-1, for example, herbal medicine, could provide a new therapeutic intervention for liver fibrosis associated with TMD in DS. (Hepatology Communications 2018;2:230-236).
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Collagen, Type IV antibody produced in mouse, clone COL-94, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Monocyte Chemotactic Protein-1 antibody produced in mouse, clone 24822, purified immunoglobulin, lyophilized powder