MilliporeSigma
  • The extracellular chaperone haptoglobin prevents serum fatty acid-promoted amyloid fibril formation of β2-microglobulin, resistance to lysosomal degradation, and cytotoxicity.
The extracellular chaperone haptoglobin prevents serum fatty acid-promoted amyloid fibril formation of β2-microglobulin, resistance to lysosomal degradation, and cytotoxicity.
The Journal of biological chemistry (2013-10-01)
Abdullah Sultan, Bakthisaran Raman, Ch Mohan Rao, Ramakrishna Tangirala
ABSTRACT
Fibril formation of β2-microglobulin and associated inflammation occur in patients on long term dialysis. We show that the plasma protein haptoglobin prevents the fatty acid-promoted de novo fibril formation of β2-microglobulin even at substoichiometric concentration. The fibrils are cytotoxic, and haptoglobin abolishes the cytotoxicity by preventing fibril formation. Haptoglobin does not alleviate the cytotoxicity of preformed fibrils. Fibrillar β2-microglobulin is resistant to lysosomal degradation. However, the species of β2-microglobulin populated in the presence of haptoglobin is susceptible to degradation. We observed that haptoglobin interacts with oligomeric prefibrillar species of β2-microglobulin but not with monomeric or fibrillar β2-microglobulin that may underlie the molecular mechanism. 1,1'-Bis(4-anilino)naphthalene-5,5'-disulfonic acid cross-linking to haptoglobin significantly compromises its chaperone activity, suggesting the involvement of hydrophobic surfaces. Haptoglobin is an acute phase protein whose level increases severalfold during inflammation, where local acidosis can occur. Our data show that haptoglobin prevents fibril formation of β2-microglobulin under conditions of physiological acidosis (between pH 5.5 and 6.5) but with relatively decreased efficiency. However, compromise in its chaperone activity under these conditions is more than compensated by its increased level of expression under inflammation. Erythrolysis is known to release hemoglobin into the plasma. Haptoglobin forms a 1:1 (mol/mol) complex with hemoglobin. This complex, like haptoglobin, interacts with the prefibrillar species of β2-microglobulin, preventing its fibril formation and the associated cytotoxicity and resistance to intracellular degradation. Thus, our study demonstrates that haptoglobin is a potential extracellular chaperone for β2-microglobulin even in moderately acidic conditions relevant during inflammation, with promising therapeutic implications in β2-microglobulin amyloid-related diseases.
Sigma-Aldrich
Anti-Human Hemoglobin antibody produced in rabbit, whole antiserum