MilliporeSigma
  • Expression of histone deacetylases in lymphoma: implication for the development of selective inhibitors.
Expression of histone deacetylases in lymphoma: implication for the development of selective inhibitors.
British journal of haematology (2009-09-25)
Annunziata Gloghini, Daniela Buglio, Noor M Khaskhely, Georgios Georgakis, Robert Z Orlowski, Sattva S Neelapu, Antonino Carbone, Anas Younes
ABSTRACT
Unselective histone deacetylase (HDAC) inhibitors are a promising novel therapy for lymphoid malignancies. However, these treatments remain empiric as the pattern of HDAC enzymes in different types of cancer, including lymphoid malignancies, remains unknown. We examined the expression of class I and class II HDACs in a panel of cell lines and tissue sections from primary lymphoid tumours. Class I enzymes were highly expressed in all cell lines and primary tumours studied, including the non-malignant reactive cells in the Hodgkin lymphoma (HL) microenvironment. The most frequently altered HDAC expression was HDAC6, as it was either weakly expressed or undetected in 9/14 (64%) of lymphoid cell lines and in 83/89 (93%) of primary lymphoma tissue specimens, including 50/52 (96%) cases of diffuse large B-cell lymphoma, and 18/22 (82%) cases of classical HL. Cell lines that had low expression level of HDAC6 demonstrated aberrant expression of hyper-acetylated tubulin, and were found to be more sensitive to the growth inhibitory effects of the class I HDAC inhibitor MGCD0103. Collectively, our data demonstrate that HDAC6 is rarely expressed in primary lymphoma cases, suggesting that it may not be an important therapeutic target in these lymphoid malignancies.
Sigma-Aldrich
Anti-Histone Deacetylase 10 (HDAC10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution