MilliporeSigma
  • Suppression of Activated FOXO Transcription Factors in the Heart Prolongs Survival in a Mouse Model of Laminopathies.
Suppression of Activated FOXO Transcription Factors in the Heart Prolongs Survival in a Mouse Model of Laminopathies.
Circulation research (2018-01-11)
Gaelle Auguste, Priyatansh Gurha, Raffaella Lombardi, Cristian Coarfa, James T Willerson, Ali J Marian
ABSTRACT
Mutations in the LMNA gene, encoding nuclear inner membrane protein lamin A/C, cause distinct phenotypes, collectively referred to as laminopathies. Heart failure, conduction defects, and arrhythmias are the common causes of death in laminopathies. The objective of this study was to identify and therapeutically target the responsible mechanism(s) for cardiac phenotype in laminopathies. Whole-heart RNA sequencing was performed before the onset of cardiac dysfunction in the Lmna-/- and matched control mice. Differentially expressed transcripts and their upstream regulators were identified, validated, and targeted by adeno-associated virus serotype 9-short hairpin RNA constructs. A total of 576 transcripts were upregulated and 233 were downregulated in the Lmna-/- mouse hearts (q<0.05). Forkhead box O (FOXO) transcription factors (TFs) were the most activated while E2 factors were the most suppressed transcriptional regulators. Transcript levels of FOXO targets were also upregulated in the isolated Lmna-/- cardiac myocytes and in the myocardium of human heart failure patients. Nuclear localization of FOXO1 and 3 was increased, whereas phosphorylated (inactive) FOXO1 and 3 levels were reduced in the Lmna-/- hearts. Gene set enrichment analysis and gene ontology showed activation of apoptosis and inflammation and suppression of cell cycle, adipogenesis, and oxidative phosphorylation in the Lmna-/- hearts. Adeno-associated virus serotype 9-short hairpin RNA-mediated suppression of FOXO TFs rescued selected molecular signatures, improved apoptosis, and prolonged survival by ≈2-fold. FOXO TFs are activated and contribute to the pathogenesis of cardiac phenotype in laminopathies. Suppression of the FOXO TFs in cardiac myocytes partially rescues the phenotype and prolongs survival. The findings identify FOXO TFs as potential therapeutic targets for cardiac phenotype in laminopathies.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 32 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit