MilliporeSigma
  • Cyclic trans-phosphorylation in a homodimer as the predominant mechanism of EGFRvIII action and regulation.
Cyclic trans-phosphorylation in a homodimer as the predominant mechanism of EGFRvIII action and regulation.
Oncotarget (2018-03-02)
Wojciech Stec, Kamila Rosiak, Cezary Treda, Maciej Smolarz, Joanna Peciak, Marcin Pacholczyk, Anna Lenart, Dawid Grzela, Ewelina Stoczynska-Fidelus, Piotr Rieske
ABSTRACT
Despite intensive research no therapies targeted against the oncogenic EGFRvIII are present in the clinic. One of the reasons is the elusive nature of the molecular structure and activity of the truncated receptor. The recent publications indicate the EGF-bound wild-type EGFR to trans-phosphorylate the EGFRvIII initiating aberrant signaling cascade. The elevated stability of the mutant receptor contributes towards oncogenic potential, preventing termination of signaling by receptor degradation. Here, we show that inhibition of phosphatases leads to a marked increase in phosphorylation of wild-type EGFR and EGFRvIII, indicating that both undergo cyclic rounds of phosphorylation and dephosphorylation on all investigated tyrosine residues, including Tyr1045. Still, we observe elevated stability of the mutant receptor, suggesting phosphorylation as insufficient to cause degradation. Hyperphosphorylation of EGFRvIII was hindered only by EGFR tyrosine kinase inhibitors. Co-immunoprecipitation as well as semi-native Western blotting structural analyses together with functional investigation of EGFRvIII's phosphorylation following depletion of wild-type EGFR by shRNA or EGF-mediated degradation indicated homodimerization as the predominant quaternary structure of the mutant receptor. Dimers were observed only under non-reducing conditions, suggesting that homodimerization is mediated by covalent bonds. Previous reports indicated cysteine at position 16 to mediate covalent homodimerization. Upon its substitution to serine, we have observed impaired formation of dimers and lower phosphorylation levels of the mutated oncogene. Based on the obtained results we propose that EGFRvIII is predominantly regulated dynamically by phosphatases that counteract the process of trans-phosphorylation occurring within the homodimers.
Sigma-Aldrich
Phenylarsine oxide, ≥97%, powder
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Sodium Orthovanadate, A broad spectrum potent inhibitor of protein tyrosine phosphatases.
Sigma-Aldrich
Anti-phospho-EGFR (Tyr1148) Antibody, Upstate®, from rabbit
Sigma-Aldrich
PhosphoDetect Anti-Phosphotyrosine Mouse mAb (PY20), liquid, clone PY20, Calbiochem®
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution