MilliporeSigma
  • Biallelic TRIP13 mutations predispose to Wilms tumor and chromosome missegregation.
Biallelic TRIP13 mutations predispose to Wilms tumor and chromosome missegregation.
Nature genetics (2017-05-30)
Shawn Yost, Bas de Wolf, Sandra Hanks, Anna Zachariou, Chiara Marcozzi, Matthew Clarke, Richarda de Voer, Banafsheh Etemad, Esther Uijttewaal, Emma Ramsay, Harriet Wylie, Anna Elliott, Susan Picton, Audrey Smith, Sarah Smithson, Sheila Seal, Elise Ruark, Gunnar Houge, Jonathan Pines, Geert J P L Kops, Nazneen Rahman
ABSTRACT
Through exome sequencing, we identified six individuals with biallelic loss-of-function mutations in TRIP13. All six developed Wilms tumor. Constitutional mosaic aneuploidies, microcephaly, developmental delay and seizures, which are features of mosaic variegated aneuploidy (MVA) syndrome, were more variably present. Through functional studies, we show that TRIP13-mutant patient cells have no detectable TRIP13 and have substantial impairment of the spindle assembly checkpoint (SAC), leading to a high rate of chromosome missegregation. Accurate segregation, as well as SAC proficiency, is rescued by restoring TRIP13 function. Individuals with biallelic TRIP13 or BUB1B mutations have a high risk of embryonal tumors, and here we show that their cells display severe SAC impairment. MVA due to biallelic CEP57 mutations, or of unknown cause, is not associated with embryonal tumors and cells from these individuals show minimal SAC deficiency. These data provide insights into the complex relationships between aneuploidy and carcinogenesis.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Mad1 Antibody, clone BB3-8, clone BB3-8, from mouse