MilliporeSigma
  • Development and characterization of six monoclonal antibodies to hemagglutinin-70 of Clostridium botulinum and their application in a sandwich ELISA.
Development and characterization of six monoclonal antibodies to hemagglutinin-70 of Clostridium botulinum and their application in a sandwich ELISA.
Monoclonal antibodies in immunodiagnosis and immunotherapy (2013-04-23)
Miles C Scotcher, Luisa W Cheng, Kathryn Ching, Jeffery McGarvey, Robert Hnasko, Larry Stanker
ABSTRACT
Botulinum neurotoxins (BoNT) are produced by Clostridium botulinum and cause severe neuroparalytic disease that if not treated quickly is often fatal. The toxin is produced as a 150 kDa precursor protein (holotoxin) that is enzymatically cleaved to form two subunits, heavy and light chains, linked by a single disulfide bond. Seven toxin serotypes are known. BoNT serotypes A1 and B1 are secreted as precursor toxic complexes (PTC) containing of the toxin and non-toxic associated proteins (NAPs) consisting of non-toxic hemagglutinin proteins (HA), designated HA17, HA34, and HA70, and a 120 kDa non-toxin non-hemagglutinin (NTNH) protein. The exact contribution of the NAPs in disease is not known, but it is thought that they protect the toxin as it passes through the harsh environment of the stomach. The structure of the complex is also poorly understood, although recent models suggest that for each molecule of toxin the PTC contains one molecule of the NTNH and multiple copies of each HA. In this paper we describe six monoclonal antibodies that specifically bind the HA70 protein found in the PTC of BoNT/A1 and /B1. Based on these antibodies, we demonstrate a rapid sandwich ELISA assay for detecting HA70.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Fc, 2.4 mg/mL, Chemicon®