MilliporeSigma
  • Identification and Profiling of a Selective and Brain Penetrant Radioligand for in Vivo Target Occupancy Measurement of Casein Kinase 1 (CK1) Inhibitors.
Identification and Profiling of a Selective and Brain Penetrant Radioligand for in Vivo Target Occupancy Measurement of Casein Kinase 1 (CK1) Inhibitors.
ACS chemical neuroscience (2017-06-14)
Travis T Wager, Paul Galatsis, Ramalakshmi Y Chandrasekaran, Todd W Butler, Jianke Li, Lei Zhang, Scot Mente, Chakrapani Subramanyam, Shenping Liu, Angela C Doran, Cheng Chang, Katherine Fisher, Sarah Grimwood, Joseph R Hedde, Michael Marconi, Klaas Schildknegt
ABSTRACT
To enable the clinical development of our CNS casein kinase 1 delta/epsilon (CK1δ/ε) inhibitor project, we investigated the possibility of developing a CNS positron emission tomography (PET) radioligand. For this effort, we focused our design and synthesis efforts on the initial CK1δ/ε inhibitor HTS hits with the goal of identifying a compound that would fulfill a set of recommended PET ligand criteria. We identified [3H]PF-5236216 (9) as a tool ligand that meets most of the key CNS PET attributes including high CNS MPO PET desirability score and kinase selectivity, CNS penetration, and low nonspecific binding. We further used [3H]-9 to determine the binding affinity for PF-670462, a literature CK1δ/ε inhibitor tool compound. Lastly, [3H]-9 was used to measure in vivo target occupancy (TO) of PF-670462 in mouse and correlated TO with CK1δ/ε in vivo pharmacology (circadian rhythm modulation).
Sigma-Aldrich
PF-05236216 hydrochloride, ≥98% (HPLC)