MilliporeSigma
  • Rapid multiresidue extraction method of organochlorinated pesticides from fish feed.
Rapid multiresidue extraction method of organochlorinated pesticides from fish feed.
Journal of chromatography. A (2004-05-01)
V Nardelli, C Palermo, D Centonze
ABSTRACT
A rapid multiresidue extraction method for organochlorinated pesticides from fish feed was developed, which is based on the extracted fat treatment by n-hexane, concentrated sulphuric acid and ENVI-carb, a graphitized non-porous carbon material. The final residue, obtained in about 50 min, was dissolved in isooctane and analysed by gas chromatography with an electron capture detector (GC/ECD). The presence of the extracted pesticides was confirmed by gas chromatography-mass spectrometry (GC/MS). Concentration of sulphuric acid and amount of ENVI-carb were optimized in order to improve analytes recovery, accuracy and detection limits. This simple and relatively fast method allowed a high recovery of the HCB, Lindane, HEPO, p,p'-DDE, p,p'-DDD, p,p'-DDT residues, with mean values in the range 68-124% at four fortification levels (12.5, 25.0, 50.0, 100.0 ng/g), and coefficients of variation between 1.9 and 20.2%. Detection limit were equal to 3.0 ng/g, related to fat, for all pesticides, and calibration curves were linear (r > 0.999) in the range of explored concentrations from the detection limit to 100 ng/g. For all pesticides a good repeatability was obtained (CV% values in the range 0.23-4.16%) when a sequence of six injections of the isooctane extraction solution was performed. The usefulness of the proposed method has been tested by the analysis of fish feed samples.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS